µltraAmp™ 384-Well NX PCR Plates, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
Designed to SBS standards with minimal warp and shrinkage after being used in thermal cycling
Certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Plates are tested for pinholes and feature a rigid skirt for gripping by robotic handlers, making them compatible with most robotics. In addition, the single cut corner allows compatibility with ABI 3730 Automated Sequencers.
Customers Who Bought This Also Bought