µltraAmp™ 96-Well Skirted PCR Plates, Sorenson BioScience
Fournisseur: Sorenson Bioscience
Certificats
A propos de cet article
Low-profile design minimizes condensation and evaporation
Certified nonpyrogenic and free of RNase, DNase, human DNA, and PCR inhibitors.
For use with MJ Research thermal cyclers and MegaBACE™ 500/1000 DNA Analysis Systems. Tight manufacturing tolerances and SBS standard dimensions make these plates ideal for automated processes. Precision-designed molds and tightly controlled processes create high-quality plates that exhibit minimal warp before and after thermal cycling. Bold alphanumeric grid. Black and white plates are designed for Real-Time PCR and other fluorescence based assays. Black plates eliminate background signal and well-to-well crosstalk. White plates deliver maximum signal amplification.
Customers Who Bought This Also Bought