384-Well PCR Plates, Sorenson BioScience
Fournisseur: Sorenson Bioscience
Certificats
A propos de cet article
Rigid skirt for gripping by robotic plate handlers
Certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Tight manufacturing tolerances and SBS standard dimensions make these plates ideal for automated processes. Precision designed molds and tightly controlled processes create high quality plates that exhibit minimal warp before and after thermal cycling. Bold alphanumeric grid. Black and white plates are designed for Real-Time PCR and other fluorescence based assays. Black plates eliminate background signal and well-to-well crosstalk. White plates deliver maximum signal amplification.
Customers Who Bought This Also Bought