µltraAmp™ 96-Well Semi-Skirted PCR Plates, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
Skirt allows robotic handling and sample labeling
Certified nonpyrogenic and free of RNase, DNase, human DNA, and PCR inhibitors.
Raised rim around perimeter for aligning purification plates. For use with Applied Biosystems thermal cyclers and DNA analyzers. Precision-designed molds and tightly controlled processes create high-quality plates that exhibit minimal warp before and after thermal cycling. Bold alphanumeric grid. Black and white plates are designed for Real-Time PCR and other fluorescence based assays. Black plates eliminate background signal and well-to-well crosstalk. White plates deliver maximum signal amplification.
Customers Who Bought This Also Bought