KEMRESIN® and KEMSHIELD® Polishes
Supplier: Kewaunee
Certificates
About this item
Apply polishes to work surfaces and shelving following detergent cleaning.
Helps keep surfaces clean, and hides minor scratches and blemishes.
Customers Who Bought This Also Bought