Mµlti™ Rigid PCR µltraPlates, 0.2 ml, Sorenson BioScience
Fournisseur: Sorenson Bioscience
Certificats
A propos de cet article
These rigid, 96-well, one-piece PCR plates are designed to eliminate plate distortion and provide a precise fit between the thermal cycler and the PCR plate
Thin-wall design reduces cycle time by allowing fast, uniform heat transfer. Grid referenced for easy identification. Complete sealing of each plate facilitates oil-free PCR and allows reaction volumes as little as 10µL. Plates can be sealed using MµlTI™ MicroMats (see 53550-032 series) and standard/pierceable CapStrips (see 53550-034 series). Certified DNase-, RNase-free. Autoclavable.
Customers Who Bought This Also Bought