Inoculum Tray and Disposable Inoculator Assembly for 96-Well Plates, Thermo Scientific
A single compartment tray, sterilized by gamma radiation designed for bacterial studies, blood toxicity and many tissue culture applications.
The disposable inoculator assembly is for use with the inoculum tray and has 95 tips and one control/blank (10 µL/tip).
Simultaneously transfers bacterial colonies from a vessel of choice to a 96-well Microtiter* format. Sterilized by gamma radiation.
Customers Who Bought This Also Bought