KEMRESIN® Countertops, Kewaunee®
# de catalogue KS-R0496-00
Fournisseur: Kewaunee
Product available on GSA Advantage®
Spécifications
- Description:Countertop
- Cat. no.:KS-R0496-00
- Supplier No.:KS-R0496-00
Spécifications
A propos de cet article
Molded epoxy resin construction provides excellent physical-, chemical-, and stain-resistance.
- Countertops are 2.5 cm (1") thick
It comes with three finished edges, including a beveled front edge; drip groove on the underside; and an attractive, black, nonglare surface.
Ordering information: Work surfaces are supplied flat or with an optional applied curb, 10.2H×1.9 cm thick (4×¾"). Please specify when ordering.
Customers Who Bought This Also Bought