Mobile Benches, Labconco®
Fournisseur: Labconco
Certificats
A propos de cet article
Create a convenient work island or add extra bench space to a wall
Note: Both benches fit fume adsorber 32948-755. Variable height bench also fits the Labconco demonstration hood.
Chemical- and heat-resistant surface. Designed to fit easily through most doorways. Heavy-duty, glacier white, epoxy coated 18-gauge steel frame is corrosion-resistant, features a centered lower support bar, and supports up to 245kg (540lbs.) while rolling over a smooth surface. Equipped with four 5.2Wx12.7dia.cm (2x5") non-marking polyurethane locking casters. Nonconductive melamine laminate work surface.
Ordering information: Manufacturer's lifetime warranty. Shipped fully assembled.
Customers Who Bought This Also Bought