Orion™ Double Junction Gel-Filled pH Electrode, Epoxy Body, Thermo Scientific
Catalogue # 14011-004
Fournisseur: Thermo Fisher Scientific
Spécifications
- Ø×L:12 ×120 mm (¹⁵/₃₂×4³/₄")
- Temperature Range:0...60 °C
- Cat. no.:14011-004
- pH Range:0 - 14
- Body Material:Epoxy
- Internal Reference:Ag/AgCl
- Junction:Wick
- Connector:BNC
- Supplier No.:9156DJWP
Spécifications
À propos de cet article
This double junction pH electrode features a durable epoxy body.
- With waterproof BNC connector
- Low-maintenance gel filling
Low maintenance, non-refillable, epoxy body pH electrode with waterproof BNC connector. For use in TRIS buffers and with proteins. Works with any pH meter with a BNC connector. Durable. Mercury-free and environmentally safe.