VWR® Contour™ Rear Scribe Filler Panel Assemblies
Supplier: VWR International
Certificates
About this item
Panels close the opening between the wall and the rear of the base cabinets at the exposed ends of the furniture assemblies.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Includes wall mounting angle.
Customers Who Bought This Also Bought