ShipSafe® Bottle Shippers, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
A propos de cet article
Lightweight, economical shippers are ideal for shipping reagents, pharmaceuticals, biological preparations, and other non-hazardous liquids in Boston round or regular bottles.
- Ideal for shipping non-hazardous liquids
- Two-piece molded EPS
- No outer carton required
- Reusable and recyclable
Lightweight, economical shippers are ideal for shipping reagents, pharmaceuticals, biological preparations, and other non-hazardous liquids in Boston round or regular bottles.
Shippers are made of two-piece molded, expanded polystyrene (EPS) foam with interlocking edges and molded-in tape grooves. No outer shipping carton is required; pressure-sensitive labels adhere directly to shipper. Shippers are reusable, recyclable, and laboratory tested and accepted for use by UPS, Parcel Post, and common carriers.
Customers Who Bought This Also Bought