A propos de cet article
Four-pack multiple convenience packs suitable for shipping hazardous materials.
Certified packaging is supplied complete with safety coated glass container, Thermoset closure with foam backed fluoropolymer resin liner, corrugated outer packaging, cushioning material, absorbent, and tape. Shipper must not make any substitutions. Failure to heed this warning would invalidate package compliance.
- Plasticoated narrow mouth bottles for liquids
- Plastic coated wide mouth bottles for solid and viscous materials
- 120 mL shipper is not plastic coated
Certified packaging is supplied complete with safety coated glass container, Thermoset closure with foam backed fluoropolymer resin liner, corrugated outer packaging, cushioning material, absorbent, and tape. Shipper must not make any substitutions. Failure to heed this warning would invalidate package compliance.
Meet UN 4G requirements (labels required).