Orion™ ISE Calibration Standards, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Take reproducible ISE measurements with Orion™ ISE Calibration Standards
For use with ion selective electrodes from ammonia to water hardness.
Ordering information: The Fluoride standard bulk pack includes 4x475mL each of 1ppm fluoride standard premixed with TISAB II and 10ppm fluoride standard premixed with TISAB II.
Packaging: All standards are supplied in 1 pint (475 mL) bottles, unless otherwise noted
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.