Mark-M-Tubes Pre-Labeled Blood Typing Tubes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These culture tubes are manufactured with a fused ceramic paint to form writing labels that will not separate from the borosilicate glass tube. Labels measure 1.9W×3.5L cm (³/₄×1³/₈").
- Pre-labeled with blood type
- Convenient package quantities
- Permanent labels placed to allow an unobstructed view of cell suspension
Certifications: Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
Frequently Bought Together