Dry Ice Infectious Substance Shipper, Qorpak®
# de catalogue 16455-262
Fournisseur: Qorpak
Small Business Enterprise
Spécifications
- Description:Dry ice infectious substance shipper
- Cat. no.:16455-262
- Supplier No.:HMS-6905M
Spécifications
A propos de cet article
Certified packaging for shipment of infectious substances.
Certified for Class 6.2 Infectious Substances. Acceptable for ground and air shipment.
- Acceptable for ground and air shipment
- Will accept up to 12 vials (vials not included)
Certified for Class 6.2 Infectious Substances.
Styrofoam core mount is suspended to accommodate dry ice. Complete with corrugated outer carton, die-cut foam, absorbent material, metal sealing ring, plastic bag, and tape. Warning labels included.
Customers Who Bought This Also Bought