Insulated Shippers, Fiberboard, Sonoco ThermoSafe
Fournisseur: Avantor
Certificats
A propos de cet article
Foam shipping container or dry ice keeper with fiberboard outer carton is ideal for mailing and laboratory use of frozen and refrigerated samples.
- Ideal for mailing frozen or refrigerated items
- Passes bursting test
- Insulated interior acts as shock absorber
Foam shipping container or dry ice keeper with fiberboard outer carton is ideal for mailing and laboratory use of frozen and refrigerated samples.
Laminated fiberboard exterior with polystyrene foam insulating interion acts as a shock absorber. Tight-fitting plug lid included.
Certifications: Passes 124 kg [275 lb] bursting test.
Customers Who Bought This Also Bought