VWR® Contour™ ADA Apron Leg Assembly
Fournisseur: VWR International
Certificats
A propos de cet article
ADA “H” Leg is made to comply with ADA requirements.
- Contour casework is manufactured of prime grade, cold-rolled, U.S. Standard sheet steel
- No ragged edges
- Light neutral color
The complete apron is used to create a knee space for a desk area or to create a free-standing table. Apron components are sold separately.
Customers Who Bought This Also Bought