VWR® Contour™ Pump Cart
# de catalogue CF-C1518-00
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Length:15"
- Width:18"
- Description:Stainless steel cart
- Height:6"
- Material:Steel
- Cat. no.:CF-C1518-00
Spécifications
A propos de cet article
Cart is for use with pump storage cabinets that are open to the floor.
- Wheels make maneuvering easy
Low-profile pump cart is made out of durable materials.
Customers Who Bought This Also Bought