Paint Can Shippers, UN Marked, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Specially designed combination package for paint shipments as well as an overpack for containers that have not passed internal pressure tests
Meet UN 4G requirements (labels required).
- One-gallon size is suitable for liquids or solids
- Pressure capability of 95kPa
Meet UN 4G requirements (labels required).
Includes paint can, absorbent material, outer corrugated carton, sealing ring, plastic bag, tape, and instructions.
Customers Who Bought This Also Bought