Petri Dish Labels, PetriStickers™, Diversified Biotech
Fournisseur: Diversified Biotech
Certificats
A propos de cet article
Labels can be applied to the back of a petri dish quickly, neatly, and reliably.
- Made with water-resistant ink, substrate, and adhesive
- Available in square grid and pie-cut formats
Labels remain firmly attached at temperatures from 4 to 42 °C (39 to 107 °F).
Customers Who Bought This Also Bought