A propos de cet article
Ultra-insulated containers maintain 2 to 8°C (35.6 to 46.4°F) in shipments over long time periods or in extreme weather conditions when used with refrigerant packs
Ultra-insulated containers maintain 2 to 8°C (35.6 to 46.4°F) in shipments over long time periods or in extreme weather conditions when used with refrigerant packs
Vapor-proof barrier enhances thermal effectiveness for a higher R-value (measure of thermal resistance) than other insulated containers. Containers are lightweight and one-piece molded. They come with an outer corrugate box sealed at the top with a soft foam polyurethane plug. Manufactured using a modular design that allows the user to recycle components. EPA-compliant, with no harmful CFCs. Meet or exceed the standards of all major shippers.