Nalgene® Petri Dish Rack, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Designed for holding plastic disposable petri dishes, these polycarbonate racks provide full side visibility for checking cultures during incubation
Lightweight, easy to load, safe for transporting. Used for media preparation, inoculation, incubation, and storage.
Customers Who Bought This Also Bought