Chemware® Chemfluor® Petri Dishes, PFA, Saint-Gobain
Fournisseur: Saint Gobain Performance Plastics
Certificats
A propos de cet article
Non-stick dishes are transparent and noncontaminating
Ideal for biological applications. The bottom is uniformly flat and level for even viewing of specimen growth. Can be repeatedly sterilized and reused. Two sizes available.
Customers Who Bought This Also Bought