Nalgene® Transparent Petri Dishes, Thermo Scientific
Catalog # 25395-350
Supplier: Thermo Fisher Scientific
Specifications
- Size:100 x 15 mm
- Cat. no.:25395-350
- Supplier No.:5500-0010
Specifications
About this item
Polymethylpentene for superior chemical and impact resistance
Top and bottom ridges interlock for stacking. Autoclavable.
Customers Who Bought This Also Bought