VWR® Culture Tubes, Disposable, with Screw-Cap Finish, Borosilicate Glass
Supplier: VWR International
Certificates
About this item
Tubes have a marking spot and reference mark for orientation during use and a threaded neck to accept screw cap.
- Borosilicate glass constuction
- Marking spot and orientation reference mark on all tubes
Please note: The capacity given below indicates the brim capacity (overflow capacity).
Delivery: Caps are not supplied, sold seperately.
Customers Who Bought This Also Bought