Drying Tube, [ST] 14/20 Joint, Wheaton, DWK Life Sciences
# de catalogue 80093-774
Fournisseur: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Spécifications
- Description:[ST] 14/20 Joint Drying Tube
- Cat. no.:80093-774
- Supplier No.:970440
Spécifications
A propos de cet article
Constructed of Wheaton 33® low extractable borosilicate glass.
The design of the tube eliminates the ball and socket connecting the scrubber and absorber components. The apparatus follows the silver diethyldithiocarbamate method described in the 18th edition of Standard Methods for the Examination of Water and Wastewater.
Certifications: Tube meets ASTM Type I Class A and USP Type I standards.
Customers Who Bought This Also Bought