U-Shaped Drying Tubes, Chemglass
Supplier: Chemglass
Certificates
About this item
Tubes feature a [ST] lower inner joint and include a rubber stopper and glass inlet tube.
Customers Who Bought This Also Bought
Tubes feature a [ST] lower inner joint and include a rubber stopper and glass inlet tube.