Imhoff Cone and Cone Racks, Wheaton, DWK Life Sciences
# de catalogue 80080-902
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Small Business Enterprise
Spécifications
- Length:45.1 cm (173/4")
- Description:Imhoff Cone
- Top Ø:10.8 cm (41/4")
- Supplier No.:W990800
Spécifications
A propos de cet article
Imhoff cone for volumetric determination of settleable solids in water and wastewater.
Styrene-acrylonitrile cone is externally graduated from plug up to 1000 ml: 0 - 1 ml in 0.1 ml; 1 - 10 ml in 0.5 ml; 10 - 40 ml in 1 ml; 40 - 100 ml in 2 ml; lines and numbers at 250, 500 and 1000 ml. Easily cleaned by removing the low-density polyethylene leak-free plug
Customers Who Bought This Also Bought