Neptune® PCR Tubes, Biotix
Fournisseur: Biotix
Certificats
A propos de cet article
These PCR tubes are made of high quality virgin polypropylene and feature uniform microthin walls for efficient heat transfer.
Tubes and caps are compatible with most leading thermal cyclers, and are autoclavable. Caps fit perfectly and create a uniform tight seal that prevents sample evaporation in the thermal cycler.
Certifications: CE certified. IVD labeled. Certified free of DNA, DNase, RNase, and endotoxins (pyrogens). Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision assuring maximum quality and compatibility. All raw materials are traceable.
Customers Who Bought This Also Bought