PCR Tubestrips and Capstrips, Sorenson BioScience
Fournisseur: Sorenson Bioscience
Certificats
A propos de cet article
Ideal for real-time PCR applications, these 8-strip tubes and caps are manufactured from optically clear polypropylene for precise and reproducible results.
- Capstrips also feature an 8-strip format, and are available with flat or domed tops
- Easily applied to tubestrips for a secure seal
- Capstrips will also fit 96-well plates and Mµlti™ µltraStrips
Uniform wall thickness ensures consistent thermal transfer, and the 8×0.2 ml tubestrip format facilitates use in most thermal cycler blocks.
Customers Who Bought This Also Bought