Spécifications
- Temperature range:0 to 100°C
- Material:Glass
- Cat. no.:CA97001-688
Spécifications
A propos de cet article
Electrode measures oxidation/reduction power of a solution
- Gold ring indicator
- Safelock™ closing ring
- ARGENTHAL™ lead-off and silver ion trap
- S7 screw head connector
Features a gold ring indicator, a Safelock™ closing ring for easy refilling and complete sealing, and an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.