Corning® Thermowell® GOLD and Thermowell PCR Tubes, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Manufactured from ultra-pure, medical grade PP suitable for RT-PCR.
- Available with flat caps for easy piercing and marking, or domed caps for efficient heat transfer
- Ultra-thin walls of uniform thickness accelerate heat transfer for shorter cycle times and higher product yields
- 100% tested for well integrity
- Certified DNase- and RNase-free and endotoxin-safe on a lot-by-lot basis
- Autoclavable at 121 °C
- Able to withstand centrifugation to 10000×g
Tubes are designed with longer cap flanges and sealed with a full-length insertion cap to safeguard against evaporation and to prevent accidental cap opening during thermal cycling.
Ordering information: Assorted pack includes the following colors: blue, green, orange, red, violet, and yellow.
Spécifications
- Catalog No:
- 29443-340
- 33502-120
- 29442-340
- 33502-122
- 33502-116
- 53495-756
- 33502-118
- 33502-124
- 33502-114
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa