VWR® PCR 12-Well Tube Strips and Caps
Fournisseur: VWR International
Certificats
A propos de cet article
Ideal for all PCR protocols, these 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
- Chemically inert
- Autoclavable
- Caps ensure a contamination-free environment
They feature smooth, thin, uniform walls for accurate thermal transfer. Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available). Caps fit precisely.
Ordering information: Tube strips with separate cap strips (53509-306) are packed with 80 tubes and 80 caps in separate bags inside a box.
Spécifications
- Catalog No:
- 53509-302
- 53509-300
- 76446-866
- 53509-306
- Environmentally Preferable:
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact