Mµlti™ PCR Tubes, Sorenson BioScience
Fournisseur: Sorenson Bioscience
Certificats
A propos de cet article
These 0.2 ml tubes fit most popular thermal cyclers.
Single polypropylene tubes with attached caps eliminate carry-over contamination and minimize evaporation. Thin-wall design ensures uniform rapid thermal transfer to reduce cycle times. Certified free of RNase, DNase, human DNA, and PCR inhibitors.
Customers Who Bought This Also Bought