Howard Leight Fusion™ Detectable Earplugs, Honeywell Safety
Fournisseur: Honeywell Safety Products
Certificats
A propos de cet article
These multiple-use earplugs use advanced manufacturing technology to deliver a revolutionary combination of all-day comfort and easy handling.
- FlexiFirm® core ensures easy insertion into the ear canal and a superior fit
- SoftFlange™ design adjusts perfectly to the ear canal to deliver unmatched comfort and superior attentuation
- Two sizes provide optimal fit for comfort and safety
- Metalized cord and stem ring offer detection for process industries
- Earplugs are stored in an included HearPack® plastic case
Customers Who Bought This Also Bought