KIMBLE® Pressure Caps for NMR Sample Tubes, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Tight-fitting caps withstand vaporization pressure buildup within the sample tube.
Caps are precision machined and available in varied polyethylene sizes.
Ordering information: Pack of assorted 10 mm caps (KT897095-1011) contains 25 each of red, black, green, and yellow.
Customers Who Bought This Also Bought