Axygen® Screw Top Microcentrifuge Tubes without Caps, Polypropylene, Corning
Supplier: Corning
Certificates
About this item
Polypropylene screw cap microcentrifuge tubes feature an alignment tab to align the tube with the centrifuge rotor for multiple applications, such as desalting.
This feature allows the tube to be re-spun without disturbing the partially pelletized sediment.
Certifications: Certified RNase-, DNase-, and pyrogen-free.
Ordering information: For compatible screw caps, see the 10011-394 series.
Customers Who Bought This Also Bought