MµlTI™ SafeSeal™ Microcentrifuge Tubes, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
These disposable microtubes are designed for use in all popular microcentrifuges.
They are manufactured from precision molded polypropylene for biotechnology applications. The cap design allows repeated openings and closings yet resists popping from internal pressure or during boiling.
Delivery: Tubes 53550-915 are supplied in individual bags of 200 of blue, yellow, violet, green, and red. Tubes 53550-778 are supplied in individual bags of 100 of blue, yellow, violet, green, and red. Tubes 53550-870 are supplied in individual bags of 80 of blue, yellow, violet, green, and red.
Frequently Bought Together