Immuno Tubes, Greiner Bio-One
# de catalogue 82050-126
Fournisseur: Greiner Bio-One
Spécifications
- Volume:5 ml
- Description:Star-Bottom Tube
- Material:Polystyrene
- Diameter ×H:12 x 75 mm
- Binding Capacity:High
- Supplier No.:115071
Spécifications
A propos de cet article
Immuno tubes are designed for all solid phase assay applications, including TSH (thyroid stimulating hormone) or other hormone determinations.
- Available in a range of binding capacities
- Polyethylene or polystyrene tube construction
- Low-binding tubes are biologically inert and have excellent chemical resistance
- Excellent optical clarity in medium- and high-binding models
Tubes are available in round- or star-bottom formats. Star-bottom tube interiors increase the available surface area to enhance levels of antigen or antibody binding. Designed for compatibility with standardized equipment, tubes are available in 4 or 5 ml volumes with a selection of low-, medium-, or high-binding capacities. Low-binding polyethylene tubes are biologically inert and offer excellent chemical resistance. Medium- and high-binding polystyrene tubes offer excellent optical clarity.
Customers Who Bought This Also Bought