KIMAX® Gas Sampling Tube with LUBRI-FLO® PTFE Stopcocks, Kimble Chase
# de catalogue 32310-176
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Capacity:250 ml (8.5 oz.)
- Length:330 mm (13")
- O.D.:50 mm (2")
- Cat. no.:32310-176
- Supplier No.:30040F-250
Spécifications
A propos de cet article
Gas sampling tube has two [ST] 4 stopcocks and tubulations on each end that accept 1 cm (³/₈") Int.Ø.
- Flexible tubing
Customers Who Bought This Also Bought