Test Tubes without Rim
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
- Tubes do not have rims
- Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Customers Who Bought This Also Bought