KIMAX® Culture Tubes, Reusable, Borosilicate Glass, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Culture tubes are made from KG-33 glass tubing and feature a fire-polished top rim and uniform wall thickness for maximum heat transfer and chemical resistance.
Each tube features a durable white ceramic enamel marking spot.
Certifications: Meet ASTM E982, Type IV requirements.
Customers Who Bought This Also Bought