VWR® Culture Tubes, Plastic, with Plug Caps, Sterile
Supplier: VWR International
Certificates
About this item
Disposable tubes feature plug-type polyethylene caps.
- Sterile
- Polystyrene or polypropylene fabrication
These tubes are available in polystyrene and polypropylene.
Ordering information: Packaged 25 per bag.
Specifications
- Catalog No:
- 60818-475
- 60818-486
- Environmentally Preferable:
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact