VWR® Round-Bottom Tubes, PTFE
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
Round-bottom tubes are designed with good temperature and chemical resistance.
- Durable pure PTFE construction
- Sturdy, heavy wall construction
- Inert and autoclavable to 280 °C (536 °F)
Isostatically molded pure, FDA approved PTFE.
Customers Who Bought This Also Bought