VWR® Conical-Bottom Centrifuge Tubes, PTFE
Supplier: VWR International
Certificates
About this item
Centrifuge tubes are constructed from sturdy PTFE and feature screw caps.
- Isostatically molded, pure, FDA-approved PTFE
- Sturdy, heavy wall construction
- Autoclavable to 280 °C (536 °F)
- Totally inert
Screw-cap design. Available in a variety of volumes and outside dimensions.
Customers Who Bought This Also Bought