VWR® Freestanding Specimen and Centrifuge Tubes, PP, with Plug Seal Caps
Supplier: VWR International
Certificates
About this item
Made of polypropylene, these tubes feature molded gradutions and a leak-proof plug seal cap designed to withstand 95 kPa of pressure.
- Polypropylene fabrication
- Skirted design
- Plug seal caps
- Available sterile or non-sterile
Skirted design enables tubes to stand upright, allowing for use without separate racks or stands.
Packaging: Tubes are available in bags of 50 with attached cap, or bulk packaged with 500 separate caps.
Specifications
- Catalog No:
- 89012-782
- 89012-778
- 89012-780
- Environmentally Preferable:
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact