Adjustable Slide Truck, Metro™
# de catalogue CA82002-468
Fournisseur: Metropolitan Wire
Spécifications
- Description:Adjustable Slide Truck
- Cat. no.:CA82002-468
- Shipping Weight:54 lbs.
- Supplier No.:APT1C-5M
Spécifications
A propos de cet article
Chrome-plated truck is designed to store and transport carriers and tote boxes
Unit is adjustable in width from 52.4 to 74.6cm (205/8 to 293/8"). Use a socket wrench to easily adjust the width. The addition of an optional grounding bracket and cable (sold separately) will make this unit conductive.
Ordering information: Truck is supplied with four 12.7cm (5") resilient tread casters (two brake and two swivel). For Olympic™ tote boxes to use with the adjustable slide truck, see 22221-298 and 82002-506 series, or contact your Avantor representative.
Customers Who Bought This Also Bought