Wintrobe Sedimentation Rate Tubes and Racks, KIMBLE CHASE®, DWK Life Sciences
Supplier: Avantor
Certificates
About this item
Wintrobe tubes are fully calibrated with yellow markings and may be filled using an aspirating bulb
Racks feature numbered stations for reliable identification and neoprene inserts that hold pipets firmly at two points to ensure pipets remain perpendicular and motionless. Built-in leveling bubble and adjusting screws assure accurate results.
Customers Who Bought This Also Bought