Caraway Capillary Tubes, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These Caraway tubes are manufactured with tapered tips for easy collection of micro blood samples.
- Designed for use with the Caraway technique
- Packed in lint-free boxes
- Ends may be sealed with clay or caps
- Non-heparinized tubes are evenly coated with ammonium heparin to prevent coagulation and do not interfere with sodium identification
Certifications: Manufactured from 90 expansion soda lime glass conforming to USP Type III requirements.
Ordering information: Sodium or lithium heparin is available as a special order.
Packaging: 600/case.
Customers Who Bought This Also Bought